accessibility menu, dialog, popup

UserName

H3CBrCH3O

CAS RN 814-75-5

IUPAC Name: 3-bromobutan-2-one
Synonyms: 3-bromo-2-butanone
Molecular Weight (g/mol): 151
Molecular Formula: C4H7BrO
InChi Key: BNBOUFHCTIFWHN-UHFFFAOYNA-N
SMILES: CC(Br)C(C)=O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)