accessibility menu, dialog, popup

UserName

HOOHOOOH

CAS RN 81-54-9

IUPAC Name: 1,2,4-trihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: purpurin purpurin, madder root hydroxylizaric acid purpurine madder root purpurin smoke brown G verantin
Molecular Weight (g/mol): 256.21
Molecular Formula: C14H8O5
InChi Key: BBNQQADTFFCFGB-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)