accessibility menu, dialog, popup

UserName

CAS RN 80221-11-0

IUPAC Name: 1-butyl-4-[2-(4-butylphenyl)ethynyl]benzene
Synonyms: 1-butyl-4-[2-(4-butylphenyl)ethynyl]benzene
Molecular Weight (g/mol): 290.45
Molecular Formula: C22H26
InChi Key: UUVMZCQRPVPWNI-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C#CC1=CC=C(CCCC)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)