accessibility menu, dialog, popup

UserName

CAS RN 800402-12-4

IUPAC Name: 2-chloro-5-iodopyridin-4-amine
Synonyms: 2-chloro-5-iodopyridin-4-amine
Molecular Weight (g/mol): 254.46
Molecular Formula: C5H4ClIN2
InChi Key: DEJUUKULVAIMNF-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=NC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results