accessibility menu, dialog, popup

UserName

CAS RN 79456-26-1

IUPAC Name: 3-chloro-5-(trifluoromethyl)pyridin-2-amine
Synonyms: 3-chloro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 196.56
Molecular Formula: C6H4ClF3N2
InChi Key: WXNPZQIRDCDLJD-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=N1)C(F)(F)F
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results