accessibility menu, dialog, popup

UserName

H3CCH3CH3HNNH

CAS RN 793-24-8

IUPAC Name: N1-(4-methylpentan-2-yl)-N4-phenylbenzene-1,4-diamine
Synonyms: santoflex 13
Molecular Weight (g/mol): 268.4
Molecular Formula: C18H24N2
InChi Key: ZZMVLMVFYMGSMY-UHFFFAOYNA-N
SMILES: CC(C)CC(C)NC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results