accessibility menu, dialog, popup

UserName

OHN(R)(S)

CAS RN 79200-56-9

IUPAC Name: (1R,4S)-2-azabicyclo[2.2.1]hept-5-en-3-one
Synonyms: (1R,4S)-2-azabicyclo[2.2.1]hept-5-en-3-one
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: DDUFYKNOXPZZIW-UHNVWZDZSA-N
SMILES: O=C1N[C@@H]2C[C@H]1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)