accessibility menu, dialog, popup

UserName

CAS RN 78351-75-4

ClHNNN

CAS RN 78351-75-4

IUPAC Name: N-(3-chlorophenyl)-4-phenylphthalazin-1-amine
Synonyms: N-(3-chlorophenyl)-4-phenylphthalazin-1-amine
Molecular Weight (g/mol): 331.8
Molecular Formula: C20H14ClN3
InChi Key: CEHQLKSLMFIHBF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(NC2=C3C=CC=CC3=C(N=N2)C2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results