accessibility menu, dialog, popup

UserName

CAS RN 78196-53-9

ONHOHNOOHNOO

CAS RN 78196-53-9

IUPAC Name: 2-{2-[(4-nitrophenyl)formamido]acetamido}acetic acid
Synonyms: {2-[(4-nitrophenyl)formamido]acetamido}acetic acid
Molecular Weight (g/mol): 281.22
Molecular Formula: C11H11N3O6
InChi Key: WQJHGFSTRONKLC-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)CNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results