missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzoylglycylglycine 99.0+%, TCI America™
Specifications
Specifications
| Purity | ≥99.0% (T) |
| CAS | 78196-53-9 |
| Color | White-Yellow |
| Format | Crystal-Powder at 20°C |
| Molecular Weight (g/mol) | 281.224 |
| Product Type | 4-Nitrobenzoylglycylglycine |
| Quantity | 1g |