accessibility menu, dialog, popup

UserName

CH3O

CAS RN 781-73-7

IUPAC Name: 1-(9H-fluoren-2-yl)ethan-1-one
Synonyms: 1-(9H-fluoren-2-yl)ethanone
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: IBASEVZORZFIIH-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2C(CC3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results