accessibility menu, dialog, popup

UserName

CAS RN 7787-47-5

IUPAC Name: beryllium(2+) dichloride
Synonyms: beryllium chloride
Molecular Weight (g/mol): 79.91
Molecular Formula: BeCl2
InChi Key: LWBPNIJBHRISSS-UHFFFAOYSA-L
SMILES: [Be++].[Cl-].[Cl-]
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results