accessibility menu, dialog, popup

UserName

CAS RN 7779-88-6

ZnONOOONOO

CAS RN 7779-88-6

IUPAC Name: zinc(2+) dinitrate
Synonyms: zinc nitrate
Molecular Weight (g/mol): 189.39
Molecular Formula: N2O6Zn
InChi Key: ONDPHDOFVYQSGI-UHFFFAOYSA-N
SMILES: [Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (9)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results