accessibility menu, dialog, popup

UserName

OHOHOOHHO

CAS RN 775-01-9

IUPAC Name: 2-(3,4-dihydroxyphenyl)-2-hydroxyacetic acid
Synonyms: dihydroxymandelic acid DHMA 3,4-dihydroxymandelic acid dihydroxymandelate 3,4-dihydroxyphenylglycolate
Molecular Weight (g/mol): 184.15
Molecular Formula: C8H8O5
InChi Key: RGHMISIYKIHAJW-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=C(O)C(O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results