accessibility menu, dialog, popup

UserName

CAS RN 7749-47-5

H3COCH3NH2NN

CAS RN 7749-47-5

IUPAC Name: 4-methoxy-6-methylpyrimidin-2-amine
Synonyms: 4-methoxy-6-methylpyrimidin-2-amine
Molecular Weight (g/mol): 139.16
Molecular Formula: C6H9N3O
InChi Key: SNWZXTZIZWBIDQ-UHFFFAOYSA-N
SMILES: COC1=CC(C)=NC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results