accessibility menu, dialog, popup

UserName

CH3OO(R)CH3Cl

CAS RN 77287-29-7

IUPAC Name: methyl (2R)-2-chloropropanoate
Synonyms: methyl (2R)-2-chloropropanoate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: JLEJCNOTNLZCHQ-GSVOUGTGSA-N
SMILES: COC(=O)[C@@H](C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results