accessibility menu, dialog, popup

UserName

NCl

CAS RN 77200-24-9

IUPAC Name: 1-(2-chloroethyl)-1H-pyrrole
Synonyms: 1-(2-chloroethyl)pyrrole
Molecular Weight (g/mol): 129.59
Molecular Formula: C6H8ClN
InChi Key: CZYATLREQUGMIQ-UHFFFAOYSA-N
SMILES: ClCCN1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results