accessibility menu, dialog, popup

UserName

CAS RN 772-59-8

H3CONBr

CAS RN 772-59-8

IUPAC Name: 2-(3-bromo-4-methoxyphenyl)acetonitrile
Synonyms: 2-(3-bromo-4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 226.07
Molecular Formula: C9H8BrNO
InChi Key: OBJKHHRZMIIEOK-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(CC#N)C=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)