accessibility menu, dialog, popup

UserName

CAS RN 77086-22-7

IUPAC Name: (1S,9R)-1-methyl-16-azatetracyclo[7.6.1.0²,⁷.0¹⁰,¹⁵]hexadeca-2,4,6,10,12,14-hexaene; (2Z)-but-2-enedioic acid
Synonyms: dizocilpine
Molecular Weight (g/mol): 337.38
Molecular Formula: C20H19NO4
InChi Key: QLTXKCWMEZIHBJ-PJGJYSAQSA-N
SMILES: OC(=O)\C=C/C(O)=O.C[C@]12N[C@H](CC3=CC=CC=C13)C1=CC=CC=C21
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)