CAS RN 77-40-7
IUPAC Name:
4-[2-(4-hydroxyphenyl)butan-2-yl]phenol
Synonyms:
bisphenol B
Molecular Weight (g/mol):
242.32
Molecular Formula:
C16H18O2
InChi Key:
HTVITOHKHWFJKO-UHFFFAOYSA-N
SMILES:
CCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
2,2-Bis(4-hydroxyphenyl)butane 98.0+%, TCI America™
CAS: 77-40-7 Molecular Formula: C16H18O2 Molecular Weight (g/mol): 242.32 MDL Number: MFCD00020179 InChI Key: HTVITOHKHWFJKO-UHFFFAOYSA-N Synonym: bisphenol b,2,2-bis 4-hydroxyphenyl butane,4,4'-butane-2,2-diyl diphenol,phenol, 4,4'-1-methylpropylidene bis,p,p'-sec-butylidenediphenol,unii-1rc731tjja,4,4'-1-methylpropylidene bisphenol,1rc731tjja,4,4'-sec-butylidenediphenol,2,2-bis p-hydroxyphenyl butane PubChem CID: 66166 ChEBI: CHEBI:34581 IUPAC Name: 4-[2-(4-hydroxyphenyl)butan-2-yl]phenol SMILES: CCC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1