accessibility menu, dialog, popup

UserName

ClOOCl

CAS RN 7693-45-0

IUPAC Name: 4-chlorophenyl carbonochloridate
Synonyms: 4-chlorophenyl carbonochloridate
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: RYWGPCLTVXMMHO-UHFFFAOYSA-N
SMILES: ClC(=O)OC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results