accessibility menu, dialog, popup

UserName

CAS RN 76280-60-9

H3COOHOOOCH3

CAS RN 76280-60-9

IUPAC Name: [3,5-bis(methoxymethoxy)phenyl]methanol
Synonyms: [3,5-bis(methoxymethoxy)phenyl]methanol
Molecular Weight (g/mol): 228.24
Molecular Formula: C11H16O5
InChi Key: QZORVGYVDDOQNT-UHFFFAOYSA-N
SMILES: COCOC1=CC(OCOC)=CC(CO)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results