accessibility menu, dialog, popup

UserName

CAS RN 76-83-5

IUPAC Name: (chlorodiphenylmethyl)benzene
Synonyms: triphenylmethyl chloride
Molecular Weight (g/mol): 278.78
Molecular Formula: C19H15Cl
InChi Key: JBWKIWSBJXDJDT-UHFFFAOYSA-N
SMILES: ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)