accessibility menu, dialog, popup

UserName

H3CONFFFCl(E/Z)

CAS RN 75999-66-5

IUPAC Name: 2,2,2-trifluoro-N-(4-methoxyphenyl)ethanecarbonimidoyl chloride
Synonyms: 2,2,2-trifluoro-N-(4-methoxyphenyl)ethanecarbonimidoyl chloride
Molecular Weight (g/mol): 237.61
Molecular Formula: C9H7ClF3NO
InChi Key: CIRVADWNFWYATJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N=C(Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results