accessibility menu, dialog, popup

UserName

CAS RN 75513-72-3

IUPAC Name: N,N'-diheptyl-N,N'-dimethylbutanediamide
Synonyms: N,N'-diheptyl-N,N'-dimethylsuccinamide
Molecular Weight (g/mol): 340.55
Molecular Formula: C20H40N2O2
InChi Key: CBEVANLIGJVSHZ-UHFFFAOYSA-N
SMILES: CCCCCCCN(C)C(=O)CCC(=O)N(C)CCCCCCC
Grade 
  • (1)
Product Line 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results