accessibility menu, dialog, popup

UserName

H3COFFFFFFFFFFFFFFF

CAS RN 754-85-8

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-one
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-pentadecafluorononan-2-one
Molecular Weight (g/mol): 412.1
Molecular Formula: C9H3F15O
InChi Key: LFMZSQGESVNRLL-UHFFFAOYSA-N
SMILES: CC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results