accessibility menu, dialog, popup

UserName

H3CNNNCH3(E/Z)

CAS RN 75232-44-9

IUPAC Name: 9-ethyl-3-[(2-methyl-2-phenylhydrazin-1-ylidene)methyl]-9H-carbazole
Synonyms: 9-ethyl-3-[(2-methyl-2-phenylhydrazin-1-ylidene)methyl]carbazole
Molecular Weight (g/mol): 327.43
Molecular Formula: C22H21N3
InChi Key: QYXUHIZLHNDFJT-UHFFFAOYSA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(C=NN(C)C1=CC=CC=C1)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results