accessibility menu, dialog, popup

UserName

IOClOOO

CAS RN 75007-13-5

IUPAC Name: diphenyliodanium perchlorate
Synonyms: diphenyliodanium perchlorate
Molecular Weight (g/mol): 380.56
Molecular Formula: C12H10ClIO4
InChi Key: JMJGNDIWCVQJJZ-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.[I+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results