accessibility menu, dialog, popup

UserName

CH3OOH3C

CAS RN 75002-34-5

IUPAC Name: 2,4-dimethylfuran-3-carbaldehyde
Synonyms: 2,4-dimethylfuran-3-carbaldehyde
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: MGSAYHDYMMHZQE-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C(C)=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)