accessibility menu, dialog, popup

UserName

HONCH2

CAS RN 75-17-2

IUPAC Name: N-methylidenehydroxylamine
Synonyms: oxime
Molecular Weight (g/mol): 45.04
Molecular Formula: CH3NO
InChi Key: SQDFHQJTAWCFIB-UHFFFAOYSA-N
SMILES: ON=C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results