accessibility menu, dialog, popup

UserName

CAS RN 749934-29-0

IUPAC Name: 2-(2-fluoro-4-methoxyphenyl)acetonitrile
Synonyms: 2-(2-fluoro-4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H8FNO
InChi Key: OKPZYRPNHFQVMZ-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results