accessibility menu, dialog, popup

UserName

CAS RN 749886-87-1

IUPAC Name: 4-methyl-N1-(3-phenylpropyl)benzene-1,2-diamine
Synonyms: 4-methyl-N1-(3-phenylpropyl)benzene-1,2-diamine
Molecular Weight (g/mol): 240.35
Molecular Formula: C16H20N2
InChi Key: YMFNPBSZFWXMAD-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(NCCCC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results