accessibility menu, dialog, popup

UserName

CH3OHNCH3O

CAS RN 7463-31-2

IUPAC Name: N-(3-acetylphenyl)acetamide
Synonyms: M-acetylaminoacetophenone
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: AFZTYHRVDOKRKV-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)C(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)