accessibility menu, dialog, popup

UserName

H3CFNCO

CAS RN 745783-74-8

IUPAC Name: 1-fluoro-4-(1-isocyanatoethyl)benzene
Synonyms: 1-fluoro-4-(1-isocyanatoethyl)benzene
Molecular Weight (g/mol): 165.17
Molecular Formula: C9H8FNO
InChi Key: MUHCZVYPBWOTTJ-UHFFFAOYNA-N
SMILES: CC(N=C=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results