accessibility menu, dialog, popup

UserName

CAS RN 74381-53-6

H3CNHONONH2NNH2NHOH3CCH3HNOH3CH3CNHOHOHNOHONHONHNHONHNHNOONHCH3OHO

CAS RN 74381-53-6

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results