missing translation for 'onlineSavingsMsg'
Learn More

Leuprorelin Acetate 97.0+%, TCI America™

Catalog No. L024925MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L024925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L024925MG Supplier TCI America Supplier No. L024925MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Purity ≥98.0% (HPLC)
CAS 74381-53-6
Color Yellow
Content And Storage Freezer (-20 °C)
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 1269.473
Product Type Leuprorelin Acetate
Quantity 25 mg