CAS RN 74-98-6
CAS RN 74-98-6
IUPAC Name:
propane
Synonyms:
propane propane (substance) petroleum gas, liquefied hydrocarbon propellant A-108 propane liquefied
Molecular Weight (g/mol):
44.1
Molecular Formula:
C3H8
InChi Key:
ATUOYWHBWRKTHZ-UHFFFAOYSA-N
SMILES:
CCC
1
–
1
of
1
results
1
Propane, SPEX CertiPrep™
CAS: 74-98-6 Molecular Formula: C3H8 Molecular Weight (g/mol): 44.097 InChI Key: ATUOYWHBWRKTHZ-UHFFFAOYSA-N PubChem CID: 6334 ChEBI: CHEBI:32879 IUPAC Name: propane SMILES: CCC