accessibility menu, dialog, popup

UserName

CH3SOOO

CAS RN 7385-85-5

IUPAC Name: naphthalen-2-yl 4-methylbenzene-1-sulfonate
Synonyms: naphthalen-2-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 298.36
Molecular Formula: C17H14O3S
InChi Key: CWCMQKWNHUXBFQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results