accessibility menu, dialog, popup

UserName

CH3SCH3H3CCH3

CAS RN 7340-90-1

IUPAC Name: (5-tert-butyl-2-methylphenyl)sulfanide
Synonyms: (5-tert-butyl-2-methylphenyl)sulfanide
Molecular Weight (g/mol): 179.3
Molecular Formula: C11H15S
InChi Key: ZSBNXOIAJFVXMP-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1[S-])C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results