accessibility menu, dialog, popup

UserName

CAS RN 7333-63-3

IUPAC Name: (4-bromobutyl)triphenylphosphanium bromide
Synonyms: (4-bromobutyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 478.21
Molecular Formula: C22H23Br2P
InChi Key: BJDNCJGRAMGIRU-UHFFFAOYSA-M
SMILES: [Br-].BrCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results