accessibility menu, dialog, popup

UserName

CAS RN 73219-89-3

IUPAC Name: 3-bromo-2,6-dimethoxybenzoate
Synonyms: 3-bromo-2,6-dimethoxybenzoate
Molecular Weight (g/mol): 260.06
Molecular Formula: C9H8BrO4
InChi Key: CUQANLQRQJHIQE-UHFFFAOYSA-M
SMILES: COC1=CC=C(Br)C(OC)=C1C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results