accessibility menu, dialog, popup

UserName

CAS RN 72773-04-7

NNNO

CAS RN 72773-04-7

IUPAC Name: 1H-1,2,3-benzotriazole-1-carbaldehyde
Synonyms: 1,2,3-benzotriazole-1-carbaldehyde
Molecular Weight (g/mol): 147.14
Molecular Formula: C7H5N3O
InChi Key: DVEIDGKSJOJJJU-UHFFFAOYSA-N
SMILES: O=CN1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results