accessibility menu, dialog, popup

UserName

CH3OOO(S)O(R)OH(S)O(S)O(R)OH(S)OH(S)OH(R)OH(R)OH(R)HO

CAS RN 72626-61-0

IUPAC Name: 7-{[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-4-methyl-2H-chromen-2-one
Synonyms: 7-{[(2S,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-4-methylchromen-2-one
Molecular Weight (g/mol): 500.45
Molecular Formula: C22H28O13
InChi Key: PRTGXBPFDYMIJH-MKQZUAMYSA-N
SMILES: CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)