accessibility menu, dialog, popup

UserName

CAS RN 7232-21-5

IUPAC Name: 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride
Synonyms: maxolon
Molecular Weight (g/mol): 336.26
Molecular Formula: C14H23Cl2N3O2
InChi Key: RVFUNJWWXKCWNS-UHFFFAOYSA-N
SMILES: Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results