accessibility menu, dialog, popup

UserName

CAS RN 72216-04-7

IUPAC Name: 2-(2-fluorophenyl)pyrrolidine
Synonyms: 2-(2-fluorophenyl)pyrrolidine
Molecular Weight (g/mol): 165.21
Molecular Formula: C10H12FN
InChi Key: NOJCVVKIBLHAGW-UHFFFAOYNA-N
SMILES: FC1=CC=CC=C1C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results