CAS RN 7215-33-0
CAS RN 7215-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diethylphosphine oxide, Thermo Scientific Chemicals
CAS: 7215-33-0 Molecular Formula: C4H11OP Molecular Weight (g/mol): 106.1