accessibility menu, dialog, popup

UserName

CAS RN 72088-94-9

IUPAC Name: 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid; 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-6-carboxylic acid
Synonyms: 5(6)-carboxyfluorescein; carboxyfluorescein
Molecular Weight (g/mol): 752.64
Molecular Formula: C42H24O14
InChi Key: BPVHBBXCESDRKW-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results