accessibility menu, dialog, popup

UserName

O

CAS RN 7206-61-3

IUPAC Name: azulene-1-carbaldehyde
Synonyms: 1-azulene carboxyaldehyde
Molecular Weight (g/mol): 156.18
Molecular Formula: C11H8O
InChi Key: CZRXLQPVJOJLML-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results