accessibility menu, dialog, popup

UserName

H3CNH3CCH3CH3I

CAS RN 7206-39-5

IUPAC Name: dimethyldioctadecylazanium iodide
Synonyms: dimethyldioctadecylazanium iodide
Molecular Weight (g/mol): 677.97
Molecular Formula: C38H80IN
InChi Key: MSSPASWTWTZPQU-UHFFFAOYSA-M
SMILES: [I-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results