accessibility menu, dialog, popup

UserName

H3CSiClClClOSiO

CAS RN 71889-02-6

IUPAC Name: silanedione; trichloro(octadecyl)silane
Synonyms: octadecyltrichlorosilane; silica
Molecular Weight (g/mol): 448.01
Molecular Formula: C18H37Cl3O2Si2
InChi Key: VYTRPUAMVCWPLO-UHFFFAOYSA-N
SMILES: O=[Si]=O.CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
Quantity 
  • (8)
  • (1)
  • (1)
Grade 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results